siRNA Details
| Gene Symbol: Alpl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287172.1 | NP_001274101.1 | 10 | 1279 | ||
| NM_001287176.1 | NP_001274105.1 | 10 | 1402 | |||
| NM_007431.3 | NP_031457.2 | 10 | 1254 | |||
| GenBank mRNA | AB473959.1 | - | 9 | 1018 | ||
| AK076459.1 | - | 10 | 1189 | |||
| AK161780.1 | - | 10 | 1190 | |||
| AK164002.1 | - | 10 | 1247 | |||
| AK166732.1 | - | 10 | 1188 | |||
| AK167023.1 | - | 10 | 1189 | |||
| BC065175.1 | - | 10 | 1177 | 6807509 | ||
| J02980.1 | - | 10 | 1193 | |||
| X13409.1 | - | 10 | 1137 | |||

Alpl
Genomic Map