siRNA Details
| Gene Symbol: Alas2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102446.1 | NP_001095916.1 | 7 | 972 | ||
| NM_009653.3 | NP_033783.1 | 7 | 1017 | |||
| GenBank mRNA | AK002642.1 | - | 7 | 988 | ||
| AK077610.1 | - | 7 | 989 | |||
| AK165257.1 | - | 7 | 980 | |||
| AK165262.1 | - | 7 | 992 | |||
| AK172094.1 | - | 7 | 981 | |||
| BC024575.1 | - | 3 | 385 | |||
| BC150870.1 | - | 7 | 964 | 9087781 | ||
| M15268.1 | - | 7 | 930 | |||
| M63244.1 | - | 7 | 987 | |||

Alas2
Genomic Map