siRNA Details
| Gene Symbol: Alk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007439.2 | NP_031465.2 | 1 | 1190 | ||
| GenBank mRNA | AK051358.1 | - | 1 | 1191 | ||
| AK158763.1 | - | 1 | 1512 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Alk| Gene Symbol: Alk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007439.2 | NP_031465.2 | 1 | 1190 | ||
| GenBank mRNA | AK051358.1 | - | 1 | 1191 | ||
| AK158763.1 | - | 1 | 1512 | |||