siRNA Details
| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007440.4 | NP_031466.2 | 3 | 421 | ||
| GenBank mRNA | AK036898.1 | - | 3 | 419 | ||
| AK036960.1 | - | 3 | 377 | |||
| AK079232.1 | - | 3 | 422 | |||
| AK087724.1 | - | 3 | 422 | |||
| AK172216.1 | - | 3 | 419 | |||
| AK172488.1 | - | 3 | 422 | |||
| BC152329.1 | - | 3 | 426 | 40141477 | ||
| BC152403.1 | - | 3 | 426 | 40141482 | ||
| S80446.1 | - | 3 | 433 | |||
| U04334.1 | - | 3 | 352 | |||

Alox12
Genomic Map