siRNA Details
| Gene Symbol: Alox12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007440.4 | NP_031466.2 | 2 | 287 | ||
| GenBank mRNA | AK036898.1 | - | 2 | 285 | ||
| AK036960.1 | - | 2 | 243 | |||
| AK079232.1 | - | 2 | 288 | |||
| AK087724.1 | - | 2 | 288 | |||
| AK172216.1 | - | 2 | 285 | |||
| AK172488.1 | - | 2 | 288 | |||
| BC152329.1 | - | 2 | 292 | 40141477 | ||
| BC152403.1 | - | 2 | 292 | 40141482 | ||
| S80446.1 | - | 2 | 299 | |||
| U04334.1 | - | 2 | 218 | |||

Alox12
Genomic Map