siRNA Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276455.1 | NP_001263384.1 | 2 | 531 | ||
| NM_054087.3 | NP_473428.1 | 2 | 531 | |||
| GenBank mRNA | AF179403.1 | - | 2 | 522 | ||
| AF216204.1 | - | 2 | 501 | |||
| AF326916.1 | - | 2 | 324 | |||
| AF360396.1 | - | 2 | 513 | |||
| AF418986.1 | - | 2 | 522 | |||
| AK044825.1 | - | 2 | 532 | |||
| BC034659.1 | - | 2 | 512 | 5359902 | ||

Slc19a2
Genomic Map