siRNA Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276455.1 | NP_001263384.1 | 2 | 585 | ||
| NM_054087.3 | NP_473428.1 | 2 | 585 | |||
| GenBank mRNA | AF179403.1 | - | 2 | 576 | ||
| AF216204.1 | - | 2 | 555 | |||
| AF326916.1 | - | 2 | 378 | |||
| AF360396.1 | - | 2 | 567 | |||
| AF418986.1 | - | 2 | 576 | |||
| AK044825.1 | - | 2 | 586 | |||
| BC034659.1 | - | 2 | 566 | 5359902 | ||

Slc19a2
Genomic Map