siRNA Details
| Gene Symbol: Slc19a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001276455.1 | NP_001263384.1 | 5 | 1444 | ||
| NM_054087.3 | NP_473428.1 | 5 | 1558 | |||
| GenBank mRNA | AF179403.1 | - | 5 | 1549 | ||
| AF216204.1 | - | 5 | 1528 | |||
| AF326916.1 | - | 5 | 1237 | |||
| AF360396.1 | - | 5 | 1540 | |||
| AF418986.1 | - | 5 | 1435 | |||
| AK044825.1 | - | 5 | 1559 | |||
| BC004021.1 | - | 1 | 112 | |||
| BC034659.1 | - | 5 | 1539 | 5359902 | ||

Slc19a2
Genomic Map