siRNA Details
| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009671.5 | NP_033801.4 | 6 | 645 | ||
| GenBank mRNA | AB011370.2 | - | 6 | 643 | ||
| AB098329.1 | - | 6 | 643 | |||
| BC139231.1 | - | 6 | 656 | 8862253 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ankfy1| Gene Symbol: Ankfy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009671.5 | NP_033801.4 | 6 | 645 | ||
| GenBank mRNA | AB011370.2 | - | 6 | 643 | ||
| AB098329.1 | - | 6 | 643 | |||
| BC139231.1 | - | 6 | 656 | 8862253 | ||