siRNA Details
| Gene Symbol: Anxa7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110794.1 | NP_001104264.1 | 8 | 934 | ||
| NM_009674.3 | NP_033804.2 | 8 | 1049 | |||
| GenBank mRNA | AK032013.1 | - | 8 | 825 | ||
| AK077579.1 | - | 8 | 742 | |||
| AK134589.1 | - | 9 | 819 | |||
| AK149654.1 | - | 8 | 741 | |||
| AK151438.1 | - | 8 | 743 | |||
| AK161613.1 | - | 8 | 743 | |||
| AK167588.1 | - | 8 | 717 | |||
| AK169199.1 | - | 8 | 716 | |||
| AK178561.1 | - | 5 | 367 | |||
| AK203311.1 | - | 3 | 187 | |||
| BC008997.1 | - | 8 | 733 | 3157926 | ||
| L13129.1 | - | 8 | 736 | |||

Anxa7
Genomic Map