siRNA Details
| Gene Symbol: Ap1m2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110300.1 | NP_001103770.1 | 9 | 1157 | ||
| NM_009678.2 | NP_033808.2 | 9 | 1151 | |||
| GenBank mRNA | AF067146.1 | - | 9 | 1125 | ||
| AK010478.1 | - | 9 | 1136 | |||
| AK082674.1 | - | 1014 | ||||
| BC003704.1 | - | 9 | 1090 | 3488231 | ||

Ap1m2
Genomic Map