siRNA Details
| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021392.4 | NP_067367.3 | 7 | 753 | ||
| GenBank mRNA | AF242858.1 | - | 7 | 621 | ||
| AK031638.1 | - | 7 | 756 | |||
| BC011174.1 | - | 7 | 638 | 4207033 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ap4m1| Gene Symbol: Ap4m1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021392.4 | NP_067367.3 | 7 | 753 | ||
| GenBank mRNA | AF242858.1 | - | 7 | 621 | ||
| AK031638.1 | - | 7 | 756 | |||
| BC011174.1 | - | 7 | 638 | 4207033 | ||