siRNA Details
Gene Symbol: Apbb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001201413.1 | NP_001188342.1 | 12 | 2188 | ||
NM_001201414.1 | NP_001188343.1 | 11 | 2191 | |||
NM_001201415.1 | NP_001188344.1 | 11 | 2125 | |||
NM_001201416.1 | NP_001188345.1 | 2 | 291 | |||
NM_009686.2 | NP_033816.1 | 13 | 2194 | |||
GenBank mRNA | AK004792.1 | - | 13 | 2048 | ||
AK153260.1 | - | 11 | 2157 | |||
AK167649.1 | - | 12 | 2053 | |||
AK169959.1 | - | 11 | 2066 | |||
AK172405.1 | - | 2 | 295 | |||
BC042797.1 | - | 3 | 333 | |||
BC076587.1 | - | 11 | 1971 | 30550087 |
