siRNA Details
| Gene Symbol: Apbb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001201413.1 | NP_001188342.1 | 15 | 2399 | ||
| NM_001201414.1 | NP_001188343.1 | 14 | 2402 | |||
| NM_001201415.1 | NP_001188344.1 | 14 | 2336 | |||
| NM_001201416.1 | NP_001188345.1 | 5 | 502 | |||
| NM_009686.2 | NP_033816.1 | 16 | 2405 | |||
| GenBank mRNA | AK004792.1 | - | 16 | 2259 | ||
| AK153260.1 | - | 14 | 2368 | |||
| AK167649.1 | - | 15 | 2264 | |||
| AK169959.1 | - | 14 | 2274 | |||
| AK172405.1 | - | 5 | 506 | |||
| BC042797.1 | - | 6 | 544 | |||
| BC076587.1 | - | 14 | 2182 | 30550087 | ||

Apbb2
Genomic Map