siRNA Details
| Gene Symbol: Aplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007467.3 | NP_031493.2 | 3 | 518 | ||
| GenBank mRNA | AK154992.1 | - | 3 | 472 | ||
| AK160871.1 | - | 3 | 470 | |||
| AK170818.1 | - | 3 | 451 | |||
| BC021877.1 | - | 3 | 497 | 4481838 | ||
| BC106158.1 | - | 3 | 425 | |||
| L04538.1 | - | 3 | 488 | |||

Aplp1
Genomic Map