siRNA Details
| Gene Symbol: Aplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007467.3 | NP_031493.2 | 4 | 610 | ||
| GenBank mRNA | AK154992.1 | - | 4 | 564 | ||
| AK160871.1 | - | 4 | 562 | |||
| AK170818.1 | - | 4 | 543 | |||
| BC021877.1 | - | 4 | 589 | 4481838 | ||
| BC106158.1 | - | 3 | 698 | |||
| L04538.1 | - | 4 | 580 | |||

Aplp1
Genomic Map