siRNA Details
| Gene Symbol: Aplp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007467.3 | NP_031493.2 | 16 | 1875 | ||
| GenBank mRNA | AK154992.1 | - | 16 | 1829 | ||
| AK160871.1 | - | 16 | 1827 | |||
| BC021877.1 | - | 16 | 1854 | 4481838 | ||
| BC106158.1 | - | 15 | 1963 | |||
| L04538.1 | - | 16 | 1845 | |||

Aplp1
Genomic Map