siRNA Details
| Gene Symbol: Aqp7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007473.4 | NP_031499.1 | 5 | 778 | ||
| GenBank mRNA | AB010100.1 | - | 5 | 669 | ||
| AB056091.1 | - | 5 | 612 | |||
| AK076642.1 | - | 5 | 746 | |||
| AK131744.1 | - | 5 | 746 | |||
| BC022223.1 | - | 3 | 408 | 4242417 | ||

Aqp7
Genomic Map