siRNA Details
| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133647.1 | - | 2522 | |||
| M37890.1 | - | 7 | 2461 | |||
| S56585.1 | - | 7 | 2540 | |||
| X53779.1 | - | 7 | 2493 | |||
| X59411.1 | - | 7 | 2492 | |||
| X59592.1 | - | 7 | 2581 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ar| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133647.1 | - | 2522 | |||
| M37890.1 | - | 7 | 2461 | |||
| S56585.1 | - | 7 | 2540 | |||
| X53779.1 | - | 7 | 2493 | |||
| X59411.1 | - | 7 | 2492 | |||
| X59592.1 | - | 7 | 2581 | |||