siRNA Details
| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133647.1 | - | 1978 | |||
| M37890.1 | - | 4 | 1917 | |||
| S56585.1 | - | 4 | 1996 | |||
| X53779.1 | - | 4 | 1949 | |||
| X59411.1 | - | 4 | 1948 | |||
| X59592.1 | - | 4 | 2037 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ar| Gene Symbol: Ar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK133647.1 | - | 1978 | |||
| M37890.1 | - | 4 | 1917 | |||
| S56585.1 | - | 4 | 1996 | |||
| X53779.1 | - | 4 | 1949 | |||
| X59411.1 | - | 4 | 1948 | |||
| X59592.1 | - | 4 | 2037 | |||