siRNA Details
| Gene Symbol: Mmp28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080453.2 | NP_536701.1 | 8 | 1604 | ||
| NM_172797.2 | NP_766385.1 | 8 | 1562 | |||
| GenBank mRNA | AK051258.1 | - | 8 | 1625 | ||
| AY065653.1 | - | 8 | 1634 | |||
| AY071828.1 | - | 8 | 1604 | |||
| AY071829.1 | - | 8 | 1562 | |||
| BC137921.1 | - | 8 | 1517 | 8860940 | ||
| BC145048.1 | - | 8 | 1589 | 9053596 | ||
| BC145049.1 | - | 8 | 1559 | 9053597 | ||

Mmp28
Genomic Map