siRNA Details
| Gene Symbol: Arg1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007482.3 | NP_031508.1 | 5 | 633 | ||
| GenBank mRNA | AK149471.1 | - | 5 | 616 | ||
| AK171417.1 | - | 4 | 467 | |||
| BC013341.1 | - | 5 | 610 | 4163626 | ||
| BC050005.1 | - | 5 | 647 | 3990246 | ||
| CB947842.1 | - | 5 | 637 | |||
| CT010173.1 | - | 5 | 531 | |||
| U51805.1 | - | 5 | 672 | |||

Arg1
Genomic Map