siRNA Details
| Gene Symbol: Arhgap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287530.1 | NP_001274459.1 | 10 | 1631 | ||
| NM_009707.4 | NP_033837.2 | 10 | 2999 | |||
| NM_178754.3 | NP_848869.1 | 10 | 1542 | |||
| GenBank mRNA | AF012273.2 | - | 2121 | |||
| AF177664.1 | - | 2121 | ||||
| AK048162.1 | - | 10 | 1496 | |||
| AK048507.1 | - | 10 | 1497 | |||
| AK077996.1 | - | 10 | 1504 | |||
| AK079651.1 | - | 10 | 1455 | |||
| AK166015.1 | - | 10 | 1462 | |||

Arhgap6
Genomic Map