siRNA Details
| Gene Symbol: Arhgap6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001287530.1 | NP_001274459.1 | 6, 7 | 1089 | ||
| NM_009707.4 | NP_033837.2 | 6, 7 | 2457 | |||
| NM_178754.3 | NP_848869.1 | 6, 7 | 1000 | |||
| GenBank mRNA | AF012273.2 | - | 1579 | |||
| AF177664.1 | - | 1579 | ||||
| AK048162.1 | - | 6, 7 | 954 | |||
| AK048507.1 | - | 6, 7 | 955 | |||
| AK077996.1 | - | 6, 7 | 962 | |||
| AK079651.1 | - | 6, 7 | 913 | |||
| AK166015.1 | - | 6, 7 | 920 | |||
| AK170575.1 | - | 6, 7 | 956 | |||

Arhgap6
Genomic Map