siRNA Details
| Gene Symbol: Serpinc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080844.4 | NP_543120.1 | 8 | 1532 | ||
| GenBank mRNA | AK050295.1 | - | 7 | 1374 | ||
| BC011165.1 | - | 7 | 1341 | |||
| BC019447.1 | - | 8 | 1492 | 5125534 | ||
| BC033377.1 | - | 7 | 1376 | 5123932 | ||
| S47225.1 | - | 7 | 1335 | |||

Serpinc1
Genomic Map