siRNA Details
| Gene Symbol: Atm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007499.2 | NP_031525.2 | 50 | 7437 | ||
| GenBank mRNA | BC067212.1 | - | 3 | 471 | ||
| BC138525.1 | - | 48 | 7330 | 8861545 | ||
| U43678.1 | - | 49 | 7339 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atm| Gene Symbol: Atm | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007499.2 | NP_031525.2 | 50 | 7437 | ||
| GenBank mRNA | BC067212.1 | - | 3 | 471 | ||
| BC138525.1 | - | 48 | 7330 | 8861545 | ||
| U43678.1 | - | 49 | 7339 | |||