siRNA Details
| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007504.2 | NP_031530.2 | 12 | 1528 | ||
| GenBank mRNA | AY081946.1 | - | 12 | 1559 | ||
| BC036292.1 | - | 12 | 1528 | 4191741 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atp2a1| Gene Symbol: Atp2a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007504.2 | NP_031530.2 | 12 | 1528 | ||
| GenBank mRNA | AY081946.1 | - | 12 | 1559 | ||
| BC036292.1 | - | 12 | 1528 | 4191741 | ||