siRNA Details
| Gene Symbol: Atp6v0d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013477.3 | NP_038505.2 | 8 | 1110 | ||
| GenBank mRNA | AK083372.1 | - | 8 | 1111 | ||
| AK133921.1 | - | 8 | 1119 | |||
| AK171515.1 | - | 8 | 1080 | |||
| BC011075.1 | - | 8 | 1001 | 3662404 | ||
| U21549.1 | - | 8 | 982 | |||

Atp6v0d1
Genomic Map