siRNA Details
Gene Symbol: Atp7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109757.2 | NP_001103227.1 | 23 | 4437 | ||
NM_009726.5 | NP_033856.3 | 23 | 4434 | |||
GenBank mRNA | AB007134.1 | - | 22 | 4273 | ||
BC141395.1 | - | 23 | 4315 | 9056008 | ||
U03434.1 | - | 23 | 4342 | |||
U03736.1 | - | 22 | 4182 | |||
U71091.1 | - | 23 | 4338 |
