siRNA Details
| Gene Symbol: Atp8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001038999.2 | NP_001034088.1 | 23 | 2220 | ||
| NM_001284345.1 | NP_001271274.1 | 23 | 2220 | |||
| NM_009727.3 | NP_033857.1 | 22 | 2175 | |||
| GenBank mRNA | AK039919.1 | - | 15 | 1991 | ||
| AK045367.1 | - | 23 | 2201 | |||
| AK076388.1 | - | 22 | 2112 | |||
| AK141559.1 | - | 23 | 2155 | |||
| BC094235.1 | - | 12 | 925 | |||
| U75321.1 | - | 22 | 1954 | |||

Atp8a1
Genomic Map