siRNA Details
| Gene Symbol: Bach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007520.2 | NP_031546.1 | 3 | 935 | ||
| GenBank mRNA | AK030871.1 | - | 3 | 885 | ||
| AK140765.1 | - | 3 | 936 | |||
| AK140903.1 | - | 3 | 936 | |||
| AK141405.1 | - | 3 | 870 | |||
| BC057894.1 | - | 3 | 856 | 4978638 | ||
| D86603.1 | - | 3 | 945 | |||

Bach1
Genomic Map