siRNA Details
| Gene Symbol: Bach2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109661.1 | NP_001103131.1 | 3 | 1898 | ||
| GenBank mRNA | BC131642.1 | - | 3 | 1806 | 40109815 | |
| Gene Symbol: Bach2os | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK144923.1 | - | 3 | 509 | ||

Bach2
Genomic Map