siRNA Details
| Gene Symbol: Bard1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007525.3 | NP_031551.1 | 4 | 595 | ||
| GenBank mRNA | AF057157.1 | - | 4 | 541 | ||
| AK145528.1 | - | 4 | 581 | |||
| BC055362.1 | - | 4 | 528 | |||
| BC128370.1 | - | 4 | 459 | 40111665 | ||
| BC128371.1 | - | 4 | 459 | 40111666 | ||

Bard1
Genomic Map