siRNA Details
Gene Symbol: Bcap29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164090.1 | NP_001157562.1 | 4, 5 | 430 | ||
NM_007530.3 | NP_031556.1 | 3, 4 | 593 | |||
GenBank mRNA | AK090068.1 | - | 4, 5 | 407 | ||
AK144754.1 | - | 4, 5 | 404 | |||
BC003303.1 | - | 3, 4 | 497 | 3584161 | ||
BC021661.1 | - | 3, 4 | 604 | 2646883 | ||
X78684.1 | - | 3, 4 | 405 |
