siRNA Details
| Gene Symbol: Bckdk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009739.3 | NP_033869.1 | 4 | 592 | ||
| GenBank mRNA | AF043070.1 | - | 3 | 433 | ||
| AK075602.1 | - | 4 | 518 | |||
| AK145479.1 | - | 4 | 563 | |||
| AK150607.1 | - | 4 | 584 | |||
| AK150723.1 | - | 4 | 584 | |||
| AK167519.1 | - | 4 | 584 | |||
| BC004077.1 | - | 1 | 20 | |||
| BC046595.1 | - | 4 | 562 | 6313106 | ||

Bckdk
Genomic Map