siRNA Details
| Gene Symbol: Bgn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007542.4 | NP_031568.2 | 2 | 269 | ||
| GenBank mRNA | AK086920.1 | - | 2 | 266 | ||
| AK135496.1 | - | 2 | 270 | |||
| AK155973.1 | - | 2 | 270 | |||
| AK164867.1 | - | 2 | 270 | |||
| AK171870.1 | - | 2 | 270 | |||
| BC005452.1 | - | 2 | 263 | 3594469 | ||
| BC019502.1 | - | 2 | 272 | 4237268 | ||
| BC052857.1 | - | 2 | 254 | 30061850 | ||
| BC057185.1 | - | 2 | 281 | |||
| L20276.1 | - | 2 | 269 | |||
| X53928.1 | - | 364 | ||||
| Y11758.1 | - | 1 | 100 | |||

Bgn
Genomic Map