siRNA Details
| Gene Symbol: Bicd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001112796.2 | NP_001106267.1 | 1 | 625 | ||
| NM_009753.4 | NP_033883.2 | 1 | 625 | |||
| GenBank mRNA | AK038585.1 | - | 1 | 540 | ||
| AK043650.1 | - | 1 | 536 | |||
| AK045975.1 | - | 1 | 544 | |||
| AK051379.1 | - | 1 | 626 | |||
| AK051718.1 | - | 1 | 559 | |||
| AK082523.1 | - | 1 | 554 | |||
| AK162699.1 | - | 1 | 725 | |||
| AK220497.1 | - | 1 | 593 | |||
| BC016192.1 | - | 1 | 328 | 4483840 | ||

Bicd1
Genomic Map