siRNA Details
Gene Symbol: Prdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK077622.1 | - | 2 | 216 | ||
AK133503.1 | - | 2 | 290 | |||
AK149344.1 | - | 2 | 181 | |||
BC129801.1 | - | 2 | 300 | |||
JX154081.1 | - | 2 | 123 | |||
U08185.1 | - | 3 | 440 |
