siRNA Details
| Gene Symbol: Blk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007549.2 | NP_031575.2 | 11 | 1556 | ||
| GenBank mRNA | AK089888.1 | - | 11 | 1556 | ||
| AK171640.1 | - | 11 | 1503 | |||
| BC030668.1 | - | 11 | 1406 | 1178763 | ||
| CT010240.1 | - | 10 | 1136 | |||
| M30903.1 | - | 11 | 1485 | |||

Blk
Genomic Map