siRNA Details
Gene Symbol: Bmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009755.3 | NP_033885.2 | 15 | 2259 | ||
GenBank mRNA | AK004995.2 | - | 15 | 1893 | ||
AK147534.1 | - | 13 | 1662 | |||
AK154239.1 | - | 15 | 2261 | |||
AK158218.1 | - | 15 | 2252 | |||
AK220295.1 | - | 15 | 2321 | |||
BC003925.1 | - | 5 | 625 | |||
BC035204.1 | - | 3 | 367 | |||
BC053923.1 | - | 15 | 2186 | |||
BC063079.1 | - | 11 | 1389 | |||
BC066062.1 | - | 15 | 2187 | 6849066 | ||
L24755.1 | - | 15 | 2036 |
