siRNA Details
| Gene Symbol: Bmp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007553.3 | NP_031579.2 | 3 | 2101 | ||
| GenBank mRNA | AK132433.1 | - | 3 | 2066 | ||
| AK133923.1 | - | 3 | 2101 | |||
| AK161862.1 | - | 3 | 1905 | |||
| BC100344.1 | - | 3 | 1061 | 30846365 | ||

Bmp2
Genomic Map