siRNA Details
| Gene Symbol: Bmpr1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009758.4 | NP_033888.2 | 10 | 1359 | ||
| GenBank mRNA | AK132051.1 | - | 10 | 1372 | ||
| AY365062.1 | - | 8 | 953 | |||
| BC042611.1 | - | 9 | 1149 | 5364272 | ||
| D16250.1 | - | 8 | 1094 | |||
| U04672.1 | - | 8 | 963 | |||
| Z23154.1 | - | 8 | 953 | |||

Bmpr1a
Genomic Map