siRNA Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007561.4 | NP_031587.1 | 12 | 2005 | ||
| GenBank mRNA | AF003942.1 | - | 12 | 2005 | ||
| AK138715.1 | - | 2 | 133 | |||
| AK148180.1 | - | 7 | 823 | |||
| AK149385.1 | - | 2886 | ||||
| U78048.1 | - | 12 | 1647 | |||

Bmpr2
Genomic Map