siRNA Details
| Gene Symbol: Bmpr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007561.4 | NP_031587.1 | 12 | 2969 | ||
| GenBank mRNA | AF003942.1 | - | 12 | 2969 | ||
| AK138715.1 | - | 2 | 1097 | |||
| AK148180.1 | - | 7 | 1787 | |||
| U78048.1 | - | 12 | 2611 | |||

Bmpr2
Genomic Map