siRNA Details
| Gene Symbol: Bnip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008238.3 | NP_001008239.1 | 4 | 507 | ||
| NM_016787.4 | NP_058067.2 | 4 | 507 | |||
| GenBank mRNA | AF035207.1 | - | 4 | 359 | ||
| AF400107.1 | - | 3 | 247 | |||
| AK014659.1 | - | 4 | 459 | |||
| AK054288.1 | - | 4 | 473 | |||
| AK161116.1 | - | 4 | 460 | |||
| AK169609.1 | - | 4 | 492 | |||
| AY591757.1 | - | 3 | 226 | |||
| AY591758.1 | - | 3 | 226 | |||
| BC012670.1 | - | 4 | 466 | 4037627 | ||

Bnip2
Genomic Map