siRNA Details
| Gene Symbol: Bub3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009774.3 | NP_033904.2 | 7 | 831 | ||
| GenBank mRNA | AF149822.1 | - | 6 | 767 | ||
| AK012744.1 | - | 2 | 35 | |||
| AK088534.1 | - | 7 | 832 | |||
| AK201702.1 | - | 1 | 25 | |||
| AK203218.1 | - | 1 | 25 | |||
| AK217256.1 | - | 1 | 18 | |||
| BC025089.1 | - | 7 | 843 | 3586753 | ||
| BC086790.1 | - | 5 | 2796 | |||
| U67327.1 | - | 7 | 818 | |||

Bub3
Genomic Map