siRNA Details
Gene Symbol: Bub3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009774.3 | NP_033904.2 | 7 | 944 | ||
GenBank mRNA | AF149822.1 | - | 6 | 880 | ||
AK012744.1 | - | 2 | 148 | |||
AK039977.1 | - | 1 | 43 | |||
AK079742.1 | - | 1 | 43 | |||
AK088534.1 | - | 7 | 945 | |||
AK180056.1 | - | 1 | 138 | |||
AK201702.1 | - | 1 | 138 | |||
AK203218.1 | - | 1 | 138 | |||
AK217256.1 | - | 1 | 131 | |||
BC025089.1 | - | 7 | 956 | 3586753 | ||
BC086790.1 | - | 5 | 2909 | |||
U67327.1 | - | 7 | 931 |
