siRNA Details
Gene Symbol: C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013484.2 | NP_038512.2 | 16 | 2111 | ||
GenBank mRNA | AK146812.1 | - | 16 | 2112 | ||
AK194662.1 | - | 1 | 18 | |||
AK203677.1 | - | 1 | 18 | |||
AK203686.1 | - | 1 | 18 | |||
BC011086.1 | - | 16 | 2074 | 3992881 | ||
M16271.1 | - | 1 | 29 | |||
M57891.1 | - | 16 | 2058 |
