siRNA Details
| Gene Symbol: C2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013484.2 | NP_038512.2 | 9, 10 | 1343 | ||
| GenBank mRNA | AK146812.1 | - | 9, 10 | 1344 | ||
| AK194054.1 | - | 2, 3 | 185 | |||
| AK211697.1 | - | 2, 3 | 204 | |||
| BC011086.1 | - | 9, 10 | 1306 | 3992881 | ||
| M57891.1 | - | 9, 10 | 1290 | |||

C2
Genomic Map