siRNA Details
| Gene Symbol: Ciita | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF042158.1 | - | 10 | 1297 | ||
| AF042159.1 | - | 11 | 1603 | |||
| AF100709.1 | - | 10 | 1480 | |||
| AF100710.1 | - | 10 | 1196 | |||
| AK040723.1 | - | 10 | 1359 | |||
| AK138822.1 | - | 10 | 1386 | |||
| AK143956.1 | - | 10 | 1468 | |||
| AK153609.1 | - | 10 | 1293 | |||
| AK155184.1 | - | 10 | 1320 | |||
| AK155311.1 | - | 10 | 1389 | |||
| U60653.1 | - | 10 | 1257 | |||

Ciita
Genomic Map